Learn More
Medchemexpress LLC N-Acetylhistamine | 673-49-4 | 153.18 | 1 ML
SDP
Catalog No. 5000436874
Encompass_Preferred
N-Acetylhistamine is a histamine metabolite that can serve as a potential biomarker for histidine metabolism in anaphylactoid reactions.
  • Histamine metabolite
  • Potential biomarker for anaphylactoid reactions
  • Human endogenous metabolite
Catalog No. 50-004-36874 Supplier Medchemexpress LLC Supplier No. HY11217510MM/1ML
N-Acetylhistamine is a histamine metabolite that can serve as a potential biomarker for histidine metabolism in anaphylactoid reactions.
  • Histamine metabolite
  • Potential biomarker for anaphylactoid reactions
  • Human endogenous metabolite