Learn More
Medchemexpress LLC N-acetylisoniazid-d4 | 1330169-81-7 | 183.20 g/mol | C8H5D4N3O2 | 1 MG
SDP
Catalog No. 5000215687
Encompass_Preferred
Deuterium-labeled acetylisoniazid provided as a research-grade reference standard for analytical and metabolic investigations. Intended for use as an internal standard in mass spectrometry, tracer experiments, and method development. Purity and recommended storage conditions are supplied in the Certificate of Analysis.
Catalog No. 50-002-15687 Supplier Medchemexpress LLC Supplier No. HY100048S1MG
Deuterium-labeled acetylisoniazid provided as a research-grade reference standard for analytical and metabolic investigations. Intended for use as an internal standard in mass spectrometry, tracer experiments, and method development. Purity and recommended storage conditions are supplied in the Certificate of Analysis.