Learn More
Cayman Chemical N-AcetylmuramIc AcId 100mg
SDP
Catalog No. 502991701
Encompass_Preferred
A component of bacterial peptidoglycan; linked to GlcNAc by β-1,4 linkages in an alternating pattern, as well as to the peptide side chains via its lactyl side chain, in native peptidoglycan
Catalog No. 50-299-1701 Supplier Cayman Chemical Supplier No. 31330100 MG
A component of bacterial peptidoglycan; linked to GlcNAc by β-1,4 linkages in an alternating pattern, as well as to the peptide side chains via its lactyl side chain, in native peptidoglycan