Learn More
Medchemexpress LLC N-Acetylmuramic acid | 10597-89-4 | 99.9% | 293.27 | 5 MG
SDP
Catalog No. 5000452629
Encompass_Preferred
N-Acetylmuramic acid is a component of the bacterial cell wall peptidoglycan, crucial for maintaining cell shape and integrity. It inhibits spore germination by targeting a coat-associated hexosaminidase and a core enzyme. This compound is required for the proliferation and maintenance of cell shape in *Bacteroides forsythus*. Additionally, N-Acetylmuramic acid has demonstrated anti-inflammatory activity by inhibiting the p38 MAPK/NF-κB signaling pathway and is orally active.
  • Destroys cell wall, enabling normal growth and typical filamentous morphology in *B. forsythus*.
  • Reduces the expression of iNOS and COX-2, and inhibits the secretion of TNF-α and IL-1β in LPS-stimulated RAW264.7 cells.
  • Inhibits *E. coli*-induced intestinal villus edema and thickening in mice, exhibiting anti-inflammatory activity.
Catalog No. 50-004-52629 Supplier Medchemexpress LLC Supplier No. HYW0092745MG
Add to Cart
Add to Cart
N-Acetylmuramic acid is a component of the bacterial cell wall peptidoglycan, crucial for maintaining cell shape and integrity. It inhibits spore germination by targeting a coat-associated hexosaminidase and a core enzyme. This compound is required for the proliferation and maintenance of cell shape in *Bacteroides forsythus*. Additionally, N-Acetylmuramic acid has demonstrated anti-inflammatory activity by inhibiting the p38 MAPK/NF-κB signaling pathway and is orally active.
  • Destroys cell wall, enabling normal growth and typical filamentous morphology in *B. forsythus*.
  • Reduces the expression of iNOS and COX-2, and inhibits the secretion of TNF-α and IL-1β in LPS-stimulated RAW264.7 cells.
  • Inhibits *E. coli*-induced intestinal villus edema and thickening in mice, exhibiting anti-inflammatory activity.