Learn More
Cayman Chemical N-Acetylmuramic Acid, Purity: ≥95%, CAS: 10597-89-4, 50 mg
SDP
Catalog No. 502990749
Encompass_Preferred
A component of bacterial peptidoglycan; linked to GlcNAc by β-1,4 linkages in an alternating pattern, as well as to the peptide side chains via its lactyl side chain, in native peptidoglycan
Catalog No. 50-299-0749 Supplier Cayman Chemical Supplier No. 3133050 MG
A component of bacterial peptidoglycan; linked to GlcNAc by β-1,4 linkages in an alternating pattern, as well as to the peptide side chains via its lactyl side chain, in native peptidoglycan