Learn More
Medchemexpress LLC N-acetylmuramyl-L-ala-γ-D-glu-meso-diaminopimelic acid | 60230-21-9 | 98.0% | 665.64 | 1 MG
SDP
Catalog No. 5000398032
Encompass_Preferred
M-TriDAP (N-Acetylmuramyl-L-Ala-γ-D-Glu-meso-diaminopimelic acid) is a NOD1/2 agonist and a biologically active peptide. It is supplied as a white to off-white solid and is intended for research use only.
  • Functions as a NOD1/2 agonist
  • Biologically active peptide
  • Appears as a white to off-white solid
  • Targeted for NOD-like receptors (NLR)
Catalog No. 50-003-98032 Supplier Medchemexpress LLC Supplier No. HYP55441MG
M-TriDAP (N-Acetylmuramyl-L-Ala-γ-D-Glu-meso-diaminopimelic acid) is a NOD1/2 agonist and a biologically active peptide. It is supplied as a white to off-white solid and is intended for research use only.
  • Functions as a NOD1/2 agonist
  • Biologically active peptide
  • Appears as a white to off-white solid
  • Targeted for NOD-like receptors (NLR)