Learn More
Medchemexpress LLC N-Acetylneuraminate lyase (CgNal) | 9027-60-5 | 5 U
SDP
Catalog No. 5000396611
Encompass_Preferred
N-Acetylneuraminate lyase (CgNal) is a class I aldolase used in biochemical studies. It catalyzes the reversible condensation of pyruvate with N-acetyl-d-mannosamine (ManNAc) to produce the sialic acid N-acetylneuraminic acid (Neu5Ac).
  • Used in biochemical studies
  • Catalyzes the reversible condensation of pyruvate with N-acetyl-d-mannosamine
  • Produces the sialic acid N-acetylneuraminic acid
  • Classified as an enzyme
  • Used in biochemical detection
  • Recombinant enzyme
  • Metabolic enzyme/protease
  • Endogenous metabolite
Catalog No. 50-003-96611 Supplier Medchemexpress LLC Supplier No. HYP29775U
N-Acetylneuraminate lyase (CgNal) is a class I aldolase used in biochemical studies. It catalyzes the reversible condensation of pyruvate with N-acetyl-d-mannosamine (ManNAc) to produce the sialic acid N-acetylneuraminic acid (Neu5Ac).
  • Used in biochemical studies
  • Catalyzes the reversible condensation of pyruvate with N-acetyl-d-mannosamine
  • Produces the sialic acid N-acetylneuraminic acid
  • Classified as an enzyme
  • Used in biochemical detection
  • Recombinant enzyme
  • Metabolic enzyme/protease
  • Endogenous metabolite