Learn More
Apexbio Technology LLC N-Acetylneuraminic acid 131-48-6 500mg
SDP
Catalog No. 5000088786
Encompass_Preferred
N-Acetylneuraminic acid (131-48-6) is a monosaccharide located at the terminal positions of glycoproteins on mammalian cell membranes It modulates cell signaling adhesion and recognition processes by altering glycoconjugate structures N-Acetylneuraminic acid exerts its biological activity primarily through regulation of sialylation patterns on surface glycoproteins which influence tumor cell proliferation adhesion dynamics and metastatic potential Key metric values such as IC50 vary significantly depending on the biological model and assay conditions Based on these properties N-Acetylneuraminic acid holds research potential in tumor biology cardiovascular injury models lipid metabolism disorders and studies of membrane protein interactions and tumor metastasis
Catalog No. 50-000-88786 Supplier Apexbio Technology LLC Supplier No. B7905500
N-Acetylneuraminic acid (131-48-6) is a monosaccharide located at the terminal positions of glycoproteins on mammalian cell membranes It modulates cell signaling adhesion and recognition processes by altering glycoconjugate structures N-Acetylneuraminic acid exerts its biological activity primarily through regulation of sialylation patterns on surface glycoproteins which influence tumor cell proliferation adhesion dynamics and metastatic potential Key metric values such as IC50 vary significantly depending on the biological model and assay conditions Based on these properties N-Acetylneuraminic acid holds research potential in tumor biology cardiovascular injury models lipid metabolism disorders and studies of membrane protein interactions and tumor metastasis