Learn More
Medchemexpress LLC N-Acetylneuraminic acid | 131-48-6 | 99.9% | 309.27 | 1 G
SDP
Catalog No. 5000428942
Encompass_Preferred
N-Acetylneuraminic acid (NANA; Lactaminic acid) is a nonphenolic sialic acid found predominately in Collocalia esculenta. It is involved in various biological processes, including myocardial injury, melanoma, and viral or bacterial infections. This compound inhibits melanogenesis by reducing tyrosinase activity and triggers myocardial injury in vitro and in vivo through activation of the Rho/Rho-associated signaling pathway. It may also prevent high fat diet-induced inflammation and oxidative stress, thereby mitigating hyperlipidemia-associated conditions.
  • Inhibits melanogenesis by reducing tyrosinase activity.
  • Triggers myocardial injury in vitro and in vivo.
  • May prevent high fat diet-induced inflammation and oxidative stress.
  • Promising for research in melanoma, coronary artery disease, obesity-related diseases, and hyperlipidemia.
Catalog No. 50-004-28942 Supplier Medchemexpress LLC Supplier No. HYI04001G
N-Acetylneuraminic acid (NANA; Lactaminic acid) is a nonphenolic sialic acid found predominately in Collocalia esculenta. It is involved in various biological processes, including myocardial injury, melanoma, and viral or bacterial infections. This compound inhibits melanogenesis by reducing tyrosinase activity and triggers myocardial injury in vitro and in vivo through activation of the Rho/Rho-associated signaling pathway. It may also prevent high fat diet-induced inflammation and oxidative stress, thereby mitigating hyperlipidemia-associated conditions.
  • Inhibits melanogenesis by reducing tyrosinase activity.
  • Triggers myocardial injury in vitro and in vivo.
  • May prevent high fat diet-induced inflammation and oxidative stress.
  • Promising for research in melanoma, coronary artery disease, obesity-related diseases, and hyperlipidemia.