Learn More
Medchemexpress LLC N-acetylneuraminic acid-13C-3 | 131-48-6 | MFCD00006620 | 99.0% | 310.26 | C10H19NO9 (13C labeled) | 10 MG
SDP
Catalog No. 5000298979
Encompass_Preferred
N-Acetylneuraminic acid-13C-3 is a stable isotope-labeled form of N-acetylneuraminic acid used as an internal standard, tracer, or reference material in analytical chemistry and metabolomics for accurate quantification and metabolic tracing.
  • 13C isotopic labeling at the 3-position for tracer studies.
  • High purity, 99.0%.
  • Suitable as an internal standard for LC-MS/MS quantification.
  • Available in small mass quantities for analytical applications.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.
Catalog No. 50-002-98979 Supplier Medchemexpress LLC Supplier No. HYI0400S310MG
N-Acetylneuraminic acid-13C-3 is a stable isotope-labeled form of N-acetylneuraminic acid used as an internal standard, tracer, or reference material in analytical chemistry and metabolomics for accurate quantification and metabolic tracing.
  • 13C isotopic labeling at the 3-position for tracer studies.
  • High purity, 99.0%.
  • Suitable as an internal standard for LC-MS/MS quantification.
  • Available in small mass quantities for analytical applications.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.