Learn More
Selleck Chemical LLC N-Acetylneuraminic acid S4792-1g
SDP
Catalog No. 503110130
Encompass_Preferred
N-Acetylneuraminic acid (Neu5Ac NeuAc Acido aceneuramico Acide aceneuramique Acidium aceneuramicum Sialic acid NANA) is the predominant sialic acid found in mammalian cells Sialic acids are negatively charged monosaccharides attached to the end of sugar chains giving rise to a wide variety of glycoproteins and glycolipids in biological fluids and cell membranes
Catalog No. 50-311-0130 Supplier Selleck Chemical LLC Supplier No. S47921G
N-Acetylneuraminic acid (Neu5Ac NeuAc Acido aceneuramico Acide aceneuramique Acidium aceneuramicum Sialic acid NANA) is the predominant sialic acid found in mammalian cells Sialic acids are negatively charged monosaccharides attached to the end of sugar chains giving rise to a wide variety of glycoproteins and glycolipids in biological fluids and cell membranes