Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylneuraminic acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012933 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2933 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2933 Supplier MilliporeSigma Supelco Supplier No. 1902310MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 131-48-6
Physical Form Powder/Crystals
Linear Formula C11H19 NO9
MDL Number MFCD00006620
Synonym 5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid; Lactaminic acid; NAN; NANA; Sialic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 309.27
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.