Learn More
Cayman Chemical N-AcetylnuramInIc AcId 100mg
SDP
Catalog No. 502973159
Encompass_Preferred
The most abundant sialic acid, found in bacteria as well as in eukaryotic cells; an essential component of human brain gangliosides and sialylated glycoproteins
Catalog No. 50-297-3159 Supplier Cayman Chemical Supplier No. 16091100 MG
The most abundant sialic acid, found in bacteria as well as in eukaryotic cells; an essential component of human brain gangliosides and sialylated glycoproteins