Learn More
Selleck Chemical LLC N-Acetylornithine
SDP
Catalog No. 5000000785
Encompass_Preferred
N-Acetylornithine is an intermediate in the enzymatic biosynthesis of the amino acid L-arginine from L-glutamate It is a minor component of deproteinized blood plasma of human blood
Catalog No. 50-000-00785 Supplier Selleck Chemical LLC Supplier No. S61165MG
N-Acetylornithine is an intermediate in the enzymatic biosynthesis of the amino acid L-arginine from L-glutamate It is a minor component of deproteinized blood plasma of human blood