Learn More
Medchemexpress LLC N-Acetylprocainamide (Standard) | 32795-44-1 | MFCD00009052 | 99.8% | 5 MG
SDP
Catalog No. 5000413485
Encompass_Preferred
N-Acetylprocainamide is a class III antiarrhythmic that blocks K+ channels.
  • Blocks K+ channels.
  • Inhibits TEA uptake in OCT2-expressing MDCK cells with an IC50 of 5 μM.
  • Inhibits TEA uptake in OCT1-expressing MDCK cells with an IC50 of 8.8 μM.
  • Decreases tensions induced by K+ and methacholine.
  • Relaxant effect is inhibited by K+ channel blockers.
  • Shows no effect on Na+ absorption or Cl- secretion.
Catalog No. 50-004-13485 Supplier Medchemexpress LLC Supplier No. HYB1109R5MG
N-Acetylprocainamide is a class III antiarrhythmic that blocks K+ channels.
  • Blocks K+ channels.
  • Inhibits TEA uptake in OCT2-expressing MDCK cells with an IC50 of 5 μM.
  • Inhibits TEA uptake in OCT1-expressing MDCK cells with an IC50 of 8.8 μM.
  • Decreases tensions induced by K+ and methacholine.
  • Relaxant effect is inhibited by K+ channel blockers.
  • Shows no effect on Na+ absorption or Cl- secretion.