Learn More
Cayman Chemical N-Acetylpuromycin, Purity: ≥98%, CAS: 22852-13-7, 5 mg
SDP
Catalog No. 502932750
Encompass_Preferred
A non-ribotoxic form of puromycin; formed in puromycin-resistant S. alboniger that endogenously express puromycin-acetyltransferase
Catalog No. 50-293-2750 Supplier Cayman Chemical Supplier No. 253355 MG
A non-ribotoxic form of puromycin; formed in puromycin-resistant S. alboniger that endogenously express puromycin-acetyltransferase