Learn More
Medchemexpress LLC N-acetylputrescine | 5699-41-2 | MFCD10024834 | 99.5% | 130.19 g/mol | C6H14N2O | 1 ML
SDP
Catalog No. 5000280478
Encompass_Preferred
N-acetylputrescine is a polyamine metabolite supplied as a ready-to-use 10 mM solution in DMSO for biochemical and biomarker research. It is suitable for metabolomics, assay development, and studies of polyamine acetylation and cell proliferation.
  • Ready-to-use 10 mM solution in DMSO.
  • 1 mL convenient volume for small-scale assays.
  • High purity: 99.5% suitable for analytical applications.
  • Reported as a biomarker in lung cancer and Parkinson's disease studies.
  • Includes CAS number for clear chemical identification.
Catalog No. 50-002-80478 Supplier Medchemexpress LLC Supplier No. HYW34260410MM/1ML
N-acetylputrescine is a polyamine metabolite supplied as a ready-to-use 10 mM solution in DMSO for biochemical and biomarker research. It is suitable for metabolomics, assay development, and studies of polyamine acetylation and cell proliferation.
  • Ready-to-use 10 mM solution in DMSO.
  • 1 mL convenient volume for small-scale assays.
  • High purity: 99.5% suitable for analytical applications.
  • Reported as a biomarker in lung cancer and Parkinson's disease studies.
  • Includes CAS number for clear chemical identification.