Learn More
Cayman Chemical N-Acetylserotonin, Purity: ≥98%, CAS: 1210-83-9, 100 mg
SDP
Catalog No. 502969758
Encompass_Preferred
The immediate precursor of melatonin that is produced from serotonin by the enzyme aralkylamine N-acetyltransferase and converted to melatonin by acetylserotonin O-methyltransferase; acts as an agonist at the melatonin receptors MT1, MT2, and MT3
Catalog No. 50-296-9758 Supplier Cayman Chemical Supplier No. 14535100 MG
The immediate precursor of melatonin that is produced from serotonin by the enzyme aralkylamine N-acetyltransferase and converted to melatonin by acetylserotonin O-methyltransferase; acts as an agonist at the melatonin receptors MT1, MT2, and MT3