Learn More
Cayman Chemical N-AcetylserotonIn 50mg
SDP
Catalog No. 502947095
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; a precursor in the synthesis of melatonin; intended for analytical forensic applications
Catalog No. 50-294-7095 Supplier Cayman Chemical Supplier No. 3368150 MG
An analytical reference standard categorized as a tryptamine; a precursor in the synthesis of melatonin; intended for analytical forensic applications