Learn More
Medchemexpress LLC N-acetyltaurine | 19213-70-8 | MFCD24549799 | 98.7% | 167.18 g/mol | C4H9NO4S | 1 ML
SDP
Catalog No. 5000284737
Encompass_Preferred
N-Acetyltaurine is an acetylated taurine derivative supplied as a research-grade reagent for biochemical and microbiological studies. Offered as a ready-to-use 10 mM solution in DMSO or as solids, it is used as a carbon and nitrogen source for microbial growth and as a substrate in enzymatic assays. Typical properties include CAS 19213-70-8, molecular formula C4H9NO4S, molecular weight 167.18 g/mol, and ~98.7% purity.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Also available as solid packs for custom preparation
  • High purity: 98.7%
  • Suitable as a microbial carbon and nitrogen source
  • Functions as an amidase substrate for enzymatic assays
  • Stable when stored under recommended conditions
Catalog No. 50-002-84737 Supplier Medchemexpress LLC Supplier No. HYW58748610MM/1ML
N-Acetyltaurine is an acetylated taurine derivative supplied as a research-grade reagent for biochemical and microbiological studies. Offered as a ready-to-use 10 mM solution in DMSO or as solids, it is used as a carbon and nitrogen source for microbial growth and as a substrate in enzymatic assays. Typical properties include CAS 19213-70-8, molecular formula C4H9NO4S, molecular weight 167.18 g/mol, and ~98.7% purity.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Also available as solid packs for custom preparation
  • High purity: 98.7%
  • Suitable as a microbial carbon and nitrogen source
  • Functions as an amidase substrate for enzymatic assays
  • Stable when stored under recommended conditions