Learn More
CPC Scientific N-Adamantanecarbonyl-DArg-Arg-Pro-Hyp-Gly-Thi-Ser-DPhe-Thi-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific BRAK006B
Encompass
Highly potent bradykinin antagonist.ONE-LETTER SEQUENCE: N-Adamantanecarbonyl-DArg-RP-Hyp-G-Thi-S-DPhe-Thi-RMOLECULAR FORMULA: C67H96N19O14S2MOLECULAR WEIGHT:1455.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-210-8856