Learn More
Medchemexpress LLC N-allyl-6-bromoquinazolin-4-amine | 307538-42-7 | MFCD02166825 | 99.9% | 264.12 | C11H10BrN3 | 200 MG
SDP
Catalog No. 5000215256
Encompass_Preferred
SMER28 is a small-molecule positive regulator of autophagy that operates via an mTOR-independent mechanism and has been reported to reduce accumulation of amyloid beta peptide. It is supplied as a research reagent in solid and solution forms for use in cellular and biochemical studies.
  • Positive regulator of autophagy via an mTOR-independent pathway.
  • Reported to prevent accumulation of amyloid beta peptide.
  • Suitable for in vitro cellular and biochemical autophagy studies.
  • Available in multiple package formats including solid and DMSO solution for assay flexibility.
  • Characterized by a defined molecular formula and molecular weight for reproducible experiments.
Catalog No. 50-002-15256 Supplier Medchemexpress LLC Supplier No. HY100200200MG
SMER28 is a small-molecule positive regulator of autophagy that operates via an mTOR-independent mechanism and has been reported to reduce accumulation of amyloid beta peptide. It is supplied as a research reagent in solid and solution forms for use in cellular and biochemical studies.
  • Positive regulator of autophagy via an mTOR-independent pathway.
  • Reported to prevent accumulation of amyloid beta peptide.
  • Suitable for in vitro cellular and biochemical autophagy studies.
  • Available in multiple package formats including solid and DMSO solution for assay flexibility.
  • Characterized by a defined molecular formula and molecular weight for reproducible experiments.