Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Allyl-N,N-Bis(Trimethylsilyl)Amine

Catalog No. 5789325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-893-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 57-893-25G Supplier Sigma Aldrich Supplier No. 5789325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Allyl-N,N-bis(trimethylsilyl)amine or N,N-bis(trimethylsilyl)prop-2-en-1-amine (AHMDS) can be used as a nucleophilic reagent to prepare: Complex bicyclic scaffolds by reacting with aryl aldehydes via imine formation followed by efficient multicomponent reactions. Dithiasuccinoyl (Dts) heterocycle by reacting with bisthiocarbamoyl chloride. Allyl selenide by reacting with aryl selenium salt in the presence of a ruthenium catalyst; Allyl selenide is utilized as a key intermediate in organic synthesis.AHMDS can also be used as a reagent in hydrosilylation and hydroboration reaction. It may be also used as an electrolyte additive.

Specifications

CAS 13454-84-7
Melting Point 250°C
Molecular Formula ClCsO4
Linear Formula CsClO4
Refractive Index n20/D 1.440
MDL Number MFCD00010956
Quantity 5 g
Molecular Weight (g/mol) 232.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.