Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-allylaniline | 589-09-3 | MFCD00008638 | 133.19 g/mol | C9H11N | 5 G
SDP

Catalog No. 5000208297
Encompass_Preferred

N-allylaniline (N-phenylallylamine) is an aromatic amine used as a reagent in chemical synthesis and biochemical assays. It is supplied in small gram quantities for laboratory research use only.

  • Aromatic primary amine reagent.
  • Molecular formula C9H11N.
  • Molecular weight 133.19 g/mol.
  • Available in gram-scale packages (1 g-100 g).
  • Intended for research use only; not for human or clinical applications.

Catalog No. 50-002-08297 Supplier Medchemexpress LLC Supplier No. HYW0870435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-allylaniline (N-phenylallylamine) is an aromatic amine used as a reagent in chemical synthesis and biochemical assays. It is supplied in small gram quantities for laboratory research use only.

  • Aromatic primary amine reagent.
  • Molecular formula C9H11N.
  • Molecular weight 133.19 g/mol.
  • Available in gram-scale packages (1 g-100 g).
  • Intended for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.