Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-allylaniline (N-phenylallylamine) | 589-09-3 | MFCD00008638 | 133.19 g/mol | C9H11N | 25 G
SDP

Catalog No. 5000209454
Encompass_Preferred

N-Allylaniline (N-phenylallylamine) is an aromatic amine reagent and intermediate used in organic synthesis and biochemical assays. It is a liquid with molecular formula C9H11N and molecular weight 133.19 g/mol, and is reported to form metal complexes and act in biochemical assay contexts. Typical handling recommends protection from light and refrigerated storage; in solvent, long-term storage at -80°C is advised.

  • Aromatic amine used as a reagent in organic synthesis
  • Molecular formula C9H11N; molecular weight 133.19 g/mol
  • Appearance: light yellow to brown liquid
  • Storage: protect from light; refrigerate for short term
  • CAS number 589-09-3 for precise identification

Catalog No. 50-002-09454 Supplier Medchemexpress LLC Supplier No. 25GHYW087043
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-Allylaniline (N-phenylallylamine) is an aromatic amine reagent and intermediate used in organic synthesis and biochemical assays. It is a liquid with molecular formula C9H11N and molecular weight 133.19 g/mol, and is reported to form metal complexes and act in biochemical assay contexts. Typical handling recommends protection from light and refrigerated storage; in solvent, long-term storage at -80°C is advised.

  • Aromatic amine used as a reagent in organic synthesis
  • Molecular formula C9H11N; molecular weight 133.19 g/mol
  • Appearance: light yellow to brown liquid
  • Storage: protect from light; refrigerate for short term
  • CAS number 589-09-3 for precise identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.