Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-allylaniline (N-phenylallylamine) | 589-09-3 | MFCD00008638 | >98.0% | 133.19 g/mol | C9H11N | 10 G
SDP

Catalog No. 5000208530
Encompass_Preferred

N-Allylaniline (N-phenylallylamine) is an aromatic amine used as a reagent in biochemical assays and laboratory research. It has the chemical formula C9H11N and a molecular weight of 133.19 g/mol. The substance is intended for research use only and is not for human or clinical applications.

  • Aromatic amine reagent
  • CAS number 589-09-3
  • Molecular formula C9H11N
  • Molecular weight 133.19 g/mol
  • available in solid quantities including 10 G
  • intended for research use only

Catalog No. 50-002-08530 Supplier Medchemexpress LLC Supplier No. HYW08704310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-Allylaniline (N-phenylallylamine) is an aromatic amine used as a reagent in biochemical assays and laboratory research. It has the chemical formula C9H11N and a molecular weight of 133.19 g/mol. The substance is intended for research use only and is not for human or clinical applications.

  • Aromatic amine reagent
  • CAS number 589-09-3
  • Molecular formula C9H11N
  • Molecular weight 133.19 g/mol
  • available in solid quantities including 10 G
  • intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.