Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(Allyloxycarbonyloxy)Succinimide

Catalog No. 73815825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-815-825G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 73-815-825G Supplier Sigma Aldrich Supplier No. 73815825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(Allyloxycarbonyloxy)succinimide (alloc-Su) can be used as: A building block for the preparation of glycopeptide scaffolds. A reagent in the synthesis of various functional cyclic carbonate monomers from 2-amino-1,3-propane diols.

Specifications

CAS 478980-01-7
Melting Point 206°C to 210°C
Molecular Formula C30H43ClN2Pd
Linear Formula C30H43ClN2Pd
Refractive Index n20/D 1.482
MDL Number MFCD07782017
Quantity 1 g
Synonym (SIPr)Pd(allyl)Cl; Allylchloro-[1,3-bis-(2,6-diisopropylphenyl)-2-imidazolidinylidene]palladium(II); CX 23
Molecular Weight (g/mol) 573.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.