Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allyloxyphthalimide 98.0+%, TCI America™
SDP

Catalog No. A144725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A144725G 25 g
A14475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A144725G Supplier TCI America Supplier No. A144725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39020-79-6
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.20
MDL Number MFCD00173307
InChI Key XVKREICBUWCANY-UHFFFAOYSA-N
PubChem CID 2765097
IUPAC Name 2-(prop-2-en-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES C=CCON1C(=O)C2=CC=CC=C2C1=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 203.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Allyloxyphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.