Learn More
AdipoGen N-Allylthiourea
SDP
Supplier:  AdipoGen CDXA0783G100
Encompass_Preferred
Chemical. CAS 109-57-9. Formula C4H8N2S. MW 116.18. Allylthiourea ATU is a strong, selective inhibitor of ammonia and nitrite oxidation. Allylthiourea is commonly used to inhibit nitritation, by targeting the ammonia monooxygenase action and chelating the copper in the active site concentration 8-80µM, ultimately hindering its function. Nitritation inhibition has been used in micropollutant biodegradability studies and in studies investigating nitritation kinetics and activity. Allylthiourea is also a potent inhibitor of methane monooxygenase MMO.
Catalog No. 50-256-1717