Learn More
Medchemexpress LLC N-alpha-(9-fluorenylmethoxycarbonyl)-4-iodo-D-phenylalanine | 205526-29-0 | MFCD00672557 | 97.0% | 513.32 g·mol⁻¹ | C24H20INO4 | 5 G
SDP
Catalog No. 5000299676
Encompass_Preferred
Fmoc-D-Phe(4-I)-OH is an Fmoc-protected derivative of D-phenylalanine bearing a para-iodo substituent. Supplied as a solid amino-acid building block, it is used in research peptide synthesis to incorporate iodinated phenylalanine residues for labeling or further functionalization.
  • Fmoc-protected amino acid for solid-phase peptide synthesis
  • Para-iodo functional group enables labeling and cross-coupling chemistry
  • Suitable for incorporation of D-phenylalanine residues in peptides
  • Typical high purity; consult lot-specific certificate of analysis
  • Powder form, store dry at recommended low temperature
Catalog No. 50-002-99676 Supplier Medchemexpress LLC Supplier No. HYW0160285G
Fmoc-D-Phe(4-I)-OH is an Fmoc-protected derivative of D-phenylalanine bearing a para-iodo substituent. Supplied as a solid amino-acid building block, it is used in research peptide synthesis to incorporate iodinated phenylalanine residues for labeling or further functionalization.
  • Fmoc-protected amino acid for solid-phase peptide synthesis
  • Para-iodo functional group enables labeling and cross-coupling chemistry
  • Suitable for incorporation of D-phenylalanine residues in peptides
  • Typical high purity; consult lot-specific certificate of analysis
  • Powder form, store dry at recommended low temperature