Learn More
Medchemexpress LLC N-alpha-Boc-D-2,3-diaminopropionic acid | 76387-70-7 | MFCD01632072 | ≥97.0% | 204.226 g/mol | C8H16N2O4 | 25 G
SDP
Catalog No. 5000303562
Encompass_Preferred
N-alpha-Boc-D-2,3-diaminopropionic acid is a Boc-protected D-amino acid derivative used as a building block in peptide synthesis and medicinal chemistry. It bears a tert-butoxycarbonyl protecting group on the alpha amino function and a free carboxylic acid, allowing selective coupling and deprotection workflows.
  • Useful building block for solid-phase and solution-phase peptide synthesis.
  • Boc protecting group facilitates orthogonal deprotection strategies.
  • Contains free carboxylic acid for standard coupling reactions.
  • Compatible with common peptide coupling reagents and solvents.
  • High purity suitable for research applications.
Catalog No. 50-003-03562 Supplier Medchemexpress LLC Supplier No. 25GHYW014692
N-alpha-Boc-D-2,3-diaminopropionic acid is a Boc-protected D-amino acid derivative used as a building block in peptide synthesis and medicinal chemistry. It bears a tert-butoxycarbonyl protecting group on the alpha amino function and a free carboxylic acid, allowing selective coupling and deprotection workflows.
  • Useful building block for solid-phase and solution-phase peptide synthesis.
  • Boc protecting group facilitates orthogonal deprotection strategies.
  • Contains free carboxylic acid for standard coupling reactions.
  • Compatible with common peptide coupling reagents and solvents.
  • High purity suitable for research applications.