Learn More
Medchemexpress LLC N-alpha-Tosyl-L-lysine chloromethyl ketone hydrochloride | 4272-74-6 | 99.0% | 369.31 | 25 MG
SDP
Catalog No. 5000438789
Encompass_Preferred
N-alpha-Tosyl-L-lysine chloromethyl ketone (TLCK) is a trypsin-like protease inhibitor that sensitizes HeLa cells to Fas-mediated cell death. It also exhibits an inhibitory effect on IFN-γ activities. This product is for research use only.
  • Trypsin-like protease inhibitor
  • Sensitizes HeLa cells to Fas-mediated cell death
  • Exhibits an inhibitory effect on IFN-γ activities
  • Soluble in H2O and DMSO at concentrations ≥ 100 mg/mL (270.78 mM)
Catalog No. 50-004-38789 Supplier Medchemexpress LLC Supplier No. 25MGHY112716
N-alpha-Tosyl-L-lysine chloromethyl ketone (TLCK) is a trypsin-like protease inhibitor that sensitizes HeLa cells to Fas-mediated cell death. It also exhibits an inhibitory effect on IFN-γ activities. This product is for research use only.
  • Trypsin-like protease inhibitor
  • Sensitizes HeLa cells to Fas-mediated cell death
  • Exhibits an inhibitory effect on IFN-γ activities
  • Soluble in H2O and DMSO at concentrations ≥ 100 mg/mL (270.78 mM)