Learn More
Medchemexpress LLC N-alpha-tosyl-L-lysine chloromethyl ketone hydrochloride | 4272-74-6 | MFCD00065395 | 99.0% | 369.31 g/mol | C14H21ClN2O3S·HCl | 5 MG
SDP
Catalog No. 5000264314
Encompass_Preferred
N-alpha-tosyl-L-lysine chloromethyl ketone hydrochloride (TLCK) is an irreversible inhibitor of trypsin and trypsin-like serine proteases. It is used in biochemical and cell biology research to suppress trypsin-like proteolytic activity, probe protease-dependent signaling, and support studies of apoptosis and related pathways.
  • Irreversible inhibitor of trypsin-like serine proteases.
  • High purity suitable for biochemical assays and analytical work.
  • Solid, white to off-white form for convenient storage and handling.
  • Commonly used in protease inhibition assays and signal-transduction studies.
  • Reported to sensitize HeLa cells to Fas-mediated cell death, useful in apoptosis research.
Catalog No. 50-002-64314 Supplier Medchemexpress LLC Supplier No. HY1127165MG
N-alpha-tosyl-L-lysine chloromethyl ketone hydrochloride (TLCK) is an irreversible inhibitor of trypsin and trypsin-like serine proteases. It is used in biochemical and cell biology research to suppress trypsin-like proteolytic activity, probe protease-dependent signaling, and support studies of apoptosis and related pathways.
  • Irreversible inhibitor of trypsin-like serine proteases.
  • High purity suitable for biochemical assays and analytical work.
  • Solid, white to off-white form for convenient storage and handling.
  • Commonly used in protease inhibition assays and signal-transduction studies.
  • Reported to sensitize HeLa cells to Fas-mediated cell death, useful in apoptosis research.