Learn More
Cayman Chemical N-Allyl-(±)-SKF 38393 (hydrobromide), Purity: ≥98%, CAS: 300561-58-4, 10 mg
SDP
Catalog No. 502946369
Encompass_Preferred
A dopamine D1 receptor partial agonist; selective for dopamine D1 over D2 receptors in radioligand binding assays (Kis = 302 and 5,250 nM, respectively); reduces food intake in rats (ED50 = 3.6 mg/kg); reduces cocaine and food self-administration in squirrel monkeys from 0.1-10 mg/kg
Catalog No. 50-294-6369 Supplier Cayman Chemical Supplier No. 3277610 MG
A dopamine D1 receptor partial agonist; selective for dopamine D1 over D2 receptors in radioligand binding assays (Kis = 302 and 5,250 nM, respectively); reduces food intake in rats (ED50 = 3.6 mg/kg); reduces cocaine and food self-administration in squirrel monkeys from 0.1-10 mg/kg