Learn More
Medchemexpress LLC N-(Amino-PEG2)-N-bis(PEG3-azide) | 2606952-86-5 | 99.85% | 550.65 | 50 MG
SDP
Catalog No. 5000351549
Encompass_Preferred
N-(Amino-PEG2)-N-bis(PEG3-azide) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-51549 Supplier Medchemexpress LLC Supplier No. HY14008750MG
N-(Amino-PEG2)-N-bis(PEG3-azide) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups