Learn More
Medchemexpress LLC N-(Amino-PEG2)-N-bis(PEG3-azide) | 2606952-86-5 | 99.9% | 550.65 | 100 MG
SDP
Catalog No. 5000351727
Encompass_Preferred
N-(Amino-PEG2)-N-bis(PEG3-azide) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Reacts with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Reacts with molecules containing DBCO or BCN groups
Catalog No. 50-003-51727 Supplier Medchemexpress LLC Supplier No. HY140087100MG
N-(Amino-PEG2)-N-bis(PEG3-azide) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Reacts with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Reacts with molecules containing DBCO or BCN groups