Learn More
Medchemexpress LLC N-(Amino-PEG3)-N-bis(PEG4-Boc) | 2353409-57-9 | 98.0% | C38H76N2O15 | 100 MG
SDP
Catalog No. 5000353008
Encompass_Preferred
N-(Amino-PEG3)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Applicable in cancer-programmed cell death research
  • Connects two different ligands for PROTAC formation
  • Targets E3 ubiquitin ligase and protein of interest
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53008 Supplier Medchemexpress LLC Supplier No. HY140254100MG
N-(Amino-PEG3)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Applicable in cancer-programmed cell death research
  • Connects two different ligands for PROTAC formation
  • Targets E3 ubiquitin ligase and protein of interest
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins