Learn More
Medchemexpress LLC N-(Amino-PEG3)-N-bis(PEG4-Boc) | 2353409-57-9 | MFCD29267156 | >98% | C38H77NO17 | 50 MG
SDP
Catalog No. 5000357236
Encompass_Preferred
N-(Amino-PEG3)-N-bis(PEG4-Boc) is a building block and PEG linker used to synthesize various PEG-based linkers. It can easily react with activated NHS esters or carboxylic acid in the presence of EDC or HATU. The t-butyl group can be deprotected under acidic conditions, and it is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Easily reacts with activated NHS esters or carboxylic acid in the presence of EDC or HATU.
  • t-butyl group can be deprotected under acidic conditions.
  • Functions as a PEG-based PROTAC linker.
Catalog No. 50-003-57236 Supplier Medchemexpress LLC Supplier No. HY14025450MG
N-(Amino-PEG3)-N-bis(PEG4-Boc) is a building block and PEG linker used to synthesize various PEG-based linkers. It can easily react with activated NHS esters or carboxylic acid in the presence of EDC or HATU. The t-butyl group can be deprotected under acidic conditions, and it is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Easily reacts with activated NHS esters or carboxylic acid in the presence of EDC or HATU.
  • t-butyl group can be deprotected under acidic conditions.
  • Functions as a PEG-based PROTAC linker.