Learn More
Medchemexpress LLC N-(Amino-PEG4)-N-bis(PEG4-Boc) | 2093153-97-8 | C40H80N2O16 | 100 MG
SDP
Catalog No. 5000357311
Encompass_Preferred
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in synthesis of protacs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57311 Supplier Medchemexpress LLC Supplier No. HY140255100MG
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in synthesis of protacs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins