Learn More
Medchemexpress LLC N-(Amino-PEG4)-N-bis(PEG4-Boc) | 2093153-97-8 | C40H80N2O16 | 250 MG
SDP
Catalog No. 5000352644
Encompass_Preferred
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two distinct ligands joined by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52644 Supplier Medchemexpress LLC Supplier No. 250MGHY140255
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two distinct ligands joined by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.