Learn More
Medchemexpress LLC N-(Amino-PEG4)-N-bis(PEG4-Boc) | 2093153-97-8 | C40H80N2O16 | 50 MG
SDP
Catalog No. 5000357323
Encompass_Preferred
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker used in the synthesis of PROTAC molecules. PROTACs are compounds designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. They achieve this by connecting two ligands: one binding to an E3 ubiquitin ligase and the other to the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTAC molecules
  • Enables selective degradation of target proteins
  • Leverages the intracellular ubiquitin-proteasome system
  • Contains two ligands connected by a linker
Catalog No. 50-003-57323 Supplier Medchemexpress LLC Supplier No. HY14025550MG
N-(Amino-PEG4)-N-bis(PEG4-Boc) is a PEG-based PROTAC linker used in the synthesis of PROTAC molecules. PROTACs are compounds designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. They achieve this by connecting two ligands: one binding to an E3 ubiquitin ligase and the other to the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTAC molecules
  • Enables selective degradation of target proteins
  • Leverages the intracellular ubiquitin-proteasome system
  • Contains two ligands connected by a linker