Learn More
Medchemexpress LLC N-(Amino-PEG5)-N-bis(PEG4-acid) | 2093152-86-2 | 99.5% | 50 MG
SDP
Catalog No. 5000338201
Encompass_Preferred
N-(Amino-PEG5)-N-bis(PEG4-acid) is a PEG derivative that contains an amino group and two terminal carboxylic acids. The amino group is reactive with carboxylic acids and activated NHS esters. The terminal carboxylic acids can react with primary amine groups in the presence of activators to form a stable amide bond. This compound is useful in the development of antibody drug conjugates (ADCs).
  • Contains an amino group and two terminal carboxylic acids
  • Amino group is reactive with carboxylic acids and activated NHS esters
  • Terminal carboxylic acids can react with primary amine groups
  • Forms a stable amide bond
  • Useful in the development of antibody drug conjugates (ADCs)
  • For research use only
Catalog No. 50-003-38201 Supplier Medchemexpress LLC Supplier No. HY13069550MG
N-(Amino-PEG5)-N-bis(PEG4-acid) is a PEG derivative that contains an amino group and two terminal carboxylic acids. The amino group is reactive with carboxylic acids and activated NHS esters. The terminal carboxylic acids can react with primary amine groups in the presence of activators to form a stable amide bond. This compound is useful in the development of antibody drug conjugates (ADCs).
  • Contains an amino group and two terminal carboxylic acids
  • Amino group is reactive with carboxylic acids and activated NHS esters
  • Terminal carboxylic acids can react with primary amine groups
  • Forms a stable amide bond
  • Useful in the development of antibody drug conjugates (ADCs)
  • For research use only