Learn More
Biomatik Corporation N Recombinant Antibody
Supplier:  Biomatik Corporation RAC07048
Encompass_Preferred
N Recombinant Antibody is a Human (Homo sapiens) Novel Coronavirus (SARS-CoV-2/ 2019-nCoV) Recombinant Antibody to N. The N Recombinant Antibody has been validated for the following applications: ELISA, WB, GICA. Host: HEK293F Cell. Species: Human (Homo sapiens) Novel Coronavirus (SARS-CoV-2/ 2019-nCoV). Immunogen: Recombinant Human Novel Coronavirus Nucleoprotein (N) (1-419Aa) (Csb-Ep3325Gmy). Applications: ELISA, WB, GICA. Target Synonyms: N. Accession Number: P0DTC9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5601