Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-arachidonoyl-gamma-aminobutyric acid | 128201-89-8 | MFCD05863981 | ≥99.0% | 389.57 | C24H39NO3 | 1 MG
SDP

Catalog No. 5000248739
Encompass_Preferred

N-Arachidonyl-GABA is a lipoamino acid related to anandamide that functions as a GABA receptor activator and has reported analgesic activity. It is provided as a high-purity solution for research applications in pharmacology and biochemical studies.

  • Cas number: 128201-89-8
  • Molecular formula: C24H39NO3
  • Molecular weight: 389.57
  • Purity: ≥99.0%
  • Form: liquid solution
  • Appearance: colorless to light yellow
  • Storage: store solution at -20°C
  • Applications: pharmacological and biochemical studies of GABAergic signaling

Catalog No. 50-002-48739 Supplier Medchemexpress LLC Supplier No. HY1035071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Arachidonyl-GABA is a lipoamino acid related to anandamide that functions as a GABA receptor activator and has reported analgesic activity. It is provided as a high-purity solution for research applications in pharmacology and biochemical studies.

  • Cas number: 128201-89-8
  • Molecular formula: C24H39NO3
  • Molecular weight: 389.57
  • Purity: ≥99.0%
  • Form: liquid solution
  • Appearance: colorless to light yellow
  • Storage: store solution at -20°C
  • Applications: pharmacological and biochemical studies of GABAergic signaling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.