Learn More
Enzo Life Sciences N-Arachidonoyl-L-phenylalanine (25mg)
SDP
Supplier:  Enzo Life Sciences BMLFA0400025
Encompass_Preferred
Putative endogenous member of a growing family of lipo-amino acids (N-acyl amino acids). Product is subject to oxidation. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-0817