Learn More
Cayman Chemical N-Arachidonoyl Dopamine, Purity: ≥98%, CAS: 199875-69-9, 50 mg
SDP
Catalog No. 502895342
Encompass_Preferred
An arachidonoyl amino acid and CB1 receptor agonist (Ki = 250 nM in rat brain membranes, which highly express CB1 receptors); selective for CB1 over CB2 receptors (Ki = 12,000 nM in rat spleen membranes, which highly express CB2 receptors); induces intracellular calcium mobilization in N18TG2 neuroblastoma cells (EC50 = 0.7 µM); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 0.25 µM); induces hypothermia, catalepsy, hypolocomotion, and analgesia in mice at 10 mg/kg
Catalog No. 50-289-5342 Supplier Cayman Chemical Supplier No. 9005750 MG
An arachidonoyl amino acid and CB1 receptor agonist (Ki = 250 nM in rat brain membranes, which highly express CB1 receptors); selective for CB1 over CB2 receptors (Ki = 12,000 nM in rat spleen membranes, which highly express CB2 receptors); induces intracellular calcium mobilization in N18TG2 neuroblastoma cells (EC50 = 0.7 µM); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 0.25 µM); induces hypothermia, catalepsy, hypolocomotion, and analgesia in mice at 10 mg/kg