Learn More
Cayman Chemical N-ArachIdonyl-L-AlnIn 10mg
SDP
Catalog No. 502895836
Encompass_Preferred
Several different arachidonoyl amino acids, including NALA, have been isolated and characterized from bovine brain. The glycine congener (NAGly) was found to suppress formalin-induced pain in rats. NALA may act at the CB receptors and/or TRPV1, but has not yet been fully characterized.
Catalog No. 50-289-5836 Supplier Cayman Chemical Supplier No. 9006510 MG
Several different arachidonoyl amino acids, including NALA, have been isolated and characterized from bovine brain. The glycine congener (NAGly) was found to suppress formalin-induced pain in rats. NALA may act at the CB receptors and/or TRPV1, but has not yet been fully characterized.