Learn More
Medchemexpress LLC N-Arachidonyl maleimide | 876305-42-9 | 98.3% | 369.54 | 1 ML
SDP
Catalog No. 5000351938
Encompass_Preferred
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It modulates the effects of 2-arachidonyl glycerol (2-AG) by increasing its endogenous levels in the brain and preventing cerebellar membrane-mediated degradation.
  • Potent and irreversible monoacylglycerol lipase (MAGL) inhibitor
  • Modulates effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • Induces hypothermia, antinociception, and catalepsy in vivo when combined with 2-AG
Catalog No. 50-003-51938 Supplier Medchemexpress LLC Supplier No. HY13656210MM/1ML
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It modulates the effects of 2-arachidonyl glycerol (2-AG) by increasing its endogenous levels in the brain and preventing cerebellar membrane-mediated degradation.
  • Potent and irreversible monoacylglycerol lipase (MAGL) inhibitor
  • Modulates effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • Induces hypothermia, antinociception, and catalepsy in vivo when combined with 2-AG