Learn More
Medchemexpress LLC N-Arachidonyl maleimide | 876305-42-9 | 98.3% | 369.54 | 100 MG
SDP
Catalog No. 5000348471
Encompass_Preferred
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It is a putative MAGL inhibitor that modulates the effects of 2-arachidonyl glycerol (2-AG), increasing endogenous levels of 2-AG in the brain and preventing cerebellar membrane-mediated degradation of 2-AG.
  • Modulates the effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • For research purposes only
Catalog No. 50-003-48471 Supplier Medchemexpress LLC Supplier No. HY136562100MG
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It is a putative MAGL inhibitor that modulates the effects of 2-arachidonyl glycerol (2-AG), increasing endogenous levels of 2-AG in the brain and preventing cerebellar membrane-mediated degradation of 2-AG.
  • Modulates the effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • For research purposes only