Learn More
Medchemexpress LLC N-Arachidonyl maleimide | 876305-42-9 | 98.3% | 369.54 | 50 MG
SDP
Catalog No. 5000348320
Encompass_Preferred
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM.
  • Modulates effects of 2-arachidonyl glycerol (2-AG) by increasing endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • Produces dose-dependent hypothermia when combined with 2-AG
  • Inhibits locomotor activity when combined with 2-AG
  • Causes antinociception when combined with 2-AG
  • Induces catalepsy when combined with 2-AG
Catalog No. 50-003-48320 Supplier Medchemexpress LLC Supplier No. HY13656250MG
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM.
  • Modulates effects of 2-arachidonyl glycerol (2-AG) by increasing endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
  • Produces dose-dependent hypothermia when combined with 2-AG
  • Inhibits locomotor activity when combined with 2-AG
  • Causes antinociception when combined with 2-AG
  • Induces catalepsy when combined with 2-AG